Chem Express X521-1 OV Badge with PrePaid Analysis for 1 Organic Vapor Pkg/5
Representative image only.
Chem Express Mfr # X525-1 – Item # WW-86378-11
Analysis performed in an AIHA-accredited lab
  • Save money and time with prepaid analysis
  • Lightweight and easy to use
  • 1 year warranty
PRODUCT OPTIONS
₹70,800.00INR / Pkg of 5
SPECS & DESCRIPTION
Specifications & Description
  • Gases MeasuredOrganic vapors
  • DescriptionOV Badge with PrePaid Analysis for 1 Organic Vapor Pkg/5
  • Warranty1 year
1 Year Warranty
Key Features
  • Save money and time with prepaid analysis
  • Lightweight and easy to use
  • 1 year warranty
More About this Item
Trace Air® Personal OV Monitor Badge has covers on each side of the monitor that can be removed to double the sampling rate. With one cover removed the monitor samples at approximately 30 mL/min, depending on the chemical. Also utilizes a new flexible, inert charcoal. Use this monitor when sampling for chemicals with a low PEL (10ppm or less), STEL monitoring or for 24 hour indoor air quality situations.
Personal monitoring badges are the most cost effective and efficient ways to monitor chemicals in your environment. Badges are easy to use and require minimal training and experience. They are small and light weight and have minimal effect on worker productivity. Badges are diffusive devices that require no additional equipment such as sampling pumps, calibration materials, or c