This item is now discontinued. We recommend the following as suggested replacements: WW-03774-39
ThermoSafe 560E Dry Ice Machine; 1-lb block; 9 to 12 blocks for European Cylinders
Representative image only.
ThermoSafe Mfr # 560E – Item # WW-03716-04
Provides up to 30% better yield from a standard 50 lb CO2 cylinder.
  • “Smart Fit” dry ice blocks offer more usable payload space in shipping containers
  • Sturdy, no-tip design and stainless steel construction
  • Easy to use and retrieve dry ice blocks, no dismantling required
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lbs)9 to 12
  • DescriptionDry Ice Machine; 1-lb block; 9 to 12 blocks for European Cylinders
  • Warranty1 year
1 Year Warranty
Key Features
  • “Smart Fit” dry ice blocks offer more usable payload space in shipping containers
  • Sturdy, no-tip design and stainless steel construction
  • Easy to use and retrieve dry ice blocks, no dismantling required
  • 1 year warranty
More About this Item
Ideal for tissue preservation, specimen shipping, and biologicals manufacturing. The latest design in bench top dry ice making, utilizes a unique heat exchange technology. No need to dismantle machine to retrieve dry ice block. Provides up to 30% better yield from a standard 50 lb (22.7 kg) CO2 cylinder (with siphon).