ThermoSafe 3730-30 Compact Insulated Shipper, 8 x 4.8 in, 1/Pk

Representative image only.
ThermoSafe Mfr # 3730-30 – Item # WW-03730-30
Ship nonhazardous materials
  • Add as many ice packs as you need — choose from a variety of sizes to meet your refrigeration needs.

PRODUCT OPTIONS

₹3,400.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Wall Thickness (cm)3.8
  • Wall Thickness (in)1 1/2
  • Inside Height (in)8
  • Outside Width (in)8
  • Outside Length (in)8
  • Outside Height (in)11
  • Inside Diameter (ID) (in)4 7/8
  • Inside Height (cm)20.3
  • Outside Width (cm)20.3
  • Outside Length (cm)20.3
  • Outside Height (cm)27.9
  • Inside Diameter (ID) (cm)12.4
  • Shipping Weight (oz)4.8
  • Shipping Weight (g)136
  • DescriptionCompact Insulated Shipper, 8 x 4.8 in, 1/Pk

Key Features

  • Add as many ice packs as you need — choose from a variety of sizes to meet your refrigeration needs.

More About this Item

Compact shippers have a cylindrical interior to hold tubes, vials, and other small items.
Note: For shipping nonhazardous materials only.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat