ThermoProbe TP9-075A-SW-SM Stainless Steel 75ft Std Probe

Representative image only.
ThermoProbe Mfr # TP9A-075-SW-SM – Item # WW-90026-05
Intrinsically safe thermometer for accurate temperature measurement in bulk liquid storage tanks
  • Ideal alternative to hazardous glass thermometers
  • Reliable and stable RTD sensor technology
  • Accuracy ±0.2°F from 32 to 200°F (0 to 100°C)
  • Easy to calibrate and make adjustments using faceplate controls
  • Cable marked every 5 feet
  • Includes 4-point NIST traceable calibration report
  • 1 year warranty

PRODUCT OPTIONS

₹340,000.00INR / Each

₹27,000.00 - ₹33,700.00INR / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Meter Measurement TypeRTD
  • Number Of Probe Channels1
  • Types Of Probes AcceptedRTD
  • Min Temperature (° F)-40
  • Max Temperature (° F)400
  • Min Temperature (° C)-40
  • Max Temperature (° C)204
  • Accuracy±0.2°F; ±0.5°F above 200°F [±0.1°C; ±0.3°C above 100°C]
  • Resolution0.01°
  • Low Battery IndicatorYes
  • Height (in)10
  • No. of Probe(s) Included1
  • Battery Life200 hours
  • Battery2 AA (included)
  • DescriptionStainless Steel 75ft Std Probe
  • Warranty1 year
1 Year WarrantyConformite EuropeenneIntrinsically Safe

Key Features

  • Ideal alternative to hazardous glass thermometers
  • Reliable and stable RTD sensor technology
  • Accuracy ±0.2°F from 32 to 200°F (0 to 100°C)
  • Easy to calibrate and make adjustments using faceplate controls
  • Cable marked every 5 feet
  • Includes 4-point NIST traceable calibration report
  • 1 year warranty

More About this Item

The ThermoProbe are designed as rugged, field-use, portable precision digital thermometers, involving volume-temperature gauging of petroleum and chemical products. Convenient design allows for easy retrieval of the cable probe and eliminates the need to bring the probe to the surface to take multiple readings. Typical applications include custody transfers, tanks, pipelines, ships, railcars, barges, tanker trucks, and proving systems.

Zone 0 intrinsically safe meters are manufactured of materials which are both immune to petrochemicals and are nonsparking. The probe assembly is constructed with a very flexible, static-dissipating, aramid fiber reinforced, nonstick cable and stainless steel components.

Large LCD displays temperature, plus arrows indicate the direction of the temperature reading and whether stability has been reached. Stabilized temperatures can be logged at numerous liquid levels for a running average and later displayed for documenting. Advanced features, accessible through a user-friendly menu, include Min/Max/Avg reading, readout integrity, photosensor-controlled backlight, selectable (°F/°C) unit of measure, battery status, and automatic shut-off.

Model TP9A features a durable, corrosion-resistant stainless steel case and a wraparound design for retrieval of the cable with probe.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat