Thermo Scientific VSTAR93 Versa Star with 2 pH/ISE, Cond, RDO/DO Modules Kit
Representative image only.
Thermo Scientific Mfr # VSTAR93 – Item # WW-58825-98
Modularity allows you to configure the meter how you need it--for now and for later
  • Customize the four measurement channels with five different parameter modules
  • Large color graphic LCD with intuitive multilanguage interface
  • Non-volatle memory holds up to 2000 data points with time and date stamp
  • Easy-to-operate USB and RS232 ports and data transfer software included
  • Up to 5 point calibration with automatic recognition for USA/NIST and DIN buffers
  • Editing calibration errors with a one point recalibration
  • Selectable cell constant for 2- or 4-cell conductivity probes
  • Selectable reading reference temperature of 5, 10, 15, 20 or 25°C
  • Choice of practical salinity or natural sea water curves for salinity
  • Accepts both polarographic and RDO(optical/luminessence) probes
  • Automatic temperature and barometic pressure compensation, manual salinity compensation
  • 3 year warranty
Include Accessories Today!
Thermo Scientific Ph/Ion Meters Accessories
9,500.00 - ₹28,500.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypeBenchtop
  • Display typeGraphic LCD
  • Buffer RecognitionDIN, NIST, and USA
  • ISE Resolution0.01 ppm
  • WaterproofYes
  • DescriptionVersa Star with 2 pH/ISE, Cond, RDO/DO Modules Kit
  • Warranty3 year
3 Year WarrantyIP54 Rated
Key Features
  • Customize the four measurement channels with five different parameter modules
  • Large color graphic LCD with intuitive multilanguage interface
  • Non-volatle memory holds up to 2000 data points with time and date stamp
  • Easy-to-operate USB and RS232 ports and data transfer software included
  • Up to 5 point calibration with automatic recognition for USA/NIST and DIN buffers
  • Editing calibration errors with a one point recalibration
  • Selectable cell constant for 2- or 4-cell conductivity probes
  • Selectable reading reference temperature of 5, 10, 15, 20 or 25°C
  • Choice of practical salinity or natural sea water curves for salinity
  • Accepts both polarographic and RDO(optical/luminessence) probes
  • Automatic temperature and barometic pressure compensation, manual salinity compensation
  • 3 year warranty
More About this Item
The VERSA STAR advanced meter provides the versatility to operate from one to four measurement channels at the same time. The five parameter modules available for the channels including; pH, pH/ISE, Conductivity, DO/RDO, and pH with LogR. Customize the meter you need with the options of meter only and modules, a meter with module package, or a full kit with meter, modules, electrode and solutions.
This versatile meter has a large color graphic LCD display. The informative display shows readings with temperature, connections, time, date, sample ID, user ID and calibration data. Plain language screen prompts, soft keys, and a multilanguage interface supports English, Spanish, French, Italian, German, and Chinese.
Non-volatle memory holds up to 2000 data sets with time and date stamp. Easy-to-operate USB and RS232 ports and data transfer software take stress out of data management.
Adds two pH/ISE, and one Conductivity, and one DO/RDO measurement modules. ROSS Triode pH/ATC electrode (05718-34); Buffer kit with 475 mL each of 4, 7, and 10 pH buffer (53350-66);DuraProbe conductivity cell (19704-70); and five 60-mL bottles of 1413 µS standard; Orion polarographic DO probe (53101-60), DO probe maintenance kit (53107-55), BOD adapters