Thermo Scientific VSTAR91 Versa Star with 2 pH/ISE, Cond, RDO/DO Modules

Representative image only.
Thermo Scientific Mfr # VSTAR91 – Item # WW-58825-97
Modularity allows you to configure the meter how you need it--for now and for later
  • Customize the four measurement channels with five different parameter modules
  • Large color graphic LCD with intuitive multilanguage interface
  • Non-volatle memory holds up to 2000 data points with time and date stamp
  • Easy-to-operate USB and RS232 ports and data transfer software included
  • Up to 5 point calibration with automatic recognition for USA/NIST and DIN buffers
  • Editing calibration errors with a one point recalibration
  • Selectable cell constant for 2- or 4-cell conductivity probes
  • Selectable reading reference temperature of 5, 10, 15, 20 or 25°C
  • Choice of practical salinity or natural sea water curves for salinity
  • Accepts both polarographic and RDO(optical/luminessence) probes
  • Automatic temperature and barometic pressure compensation, manual salinity compensation
  • 3 year warranty

PRODUCT OPTIONS

Call For Price

₹27,100.00 - ₹37,000.00INR / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Oakton Electrode Storage Solution; 500 mL
₹3,900.00INR / Pint of 1
Include Accessories Today!
Thermo Scientific Ph/Ion Meters Accessories
9,500.00 - ₹30,010.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display typeGraphic LCD
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 6 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min ISE0 to 19999 ppm
  • ISE Resolution0.01 ppm
  • ISE Accuracy±0.05% of reading
  • Conductivity Accuracy±0.5% FS +1 LSD
  • Min TDS0 to 200 ppt
  • Min Salinity0 to 80 ppt
  • Salinity Accuracy±0.1 ppt
  • Resistivity Accuracy±0.5% ±1 digit
  • Min Dissolved Oxygen0 to 90 ppm; 0 to 600 %
  • Dissolved Oxygen Accuracy±0.2 ppm
  • Min Temperature (° C)-5
  • Max Temperature (° C)105
  • Min Temperature (° F)23
  • Max Temperature (° F)221
  • InterfaceRS-232 and USB
  • WaterproofYes
  • Power (VAC)90 to 260
  • Power (Hz)50 or 60
  • Battery4 AA (not included)
  • Width (cm)17.78
  • Height (cm)24.765
  • Width (in)7
  • Length (in)3 3/8
  • Height (in)9 3/4
  • DescriptionVersa Star with 2 pH/ISE, Cond, RDO/DO Modules
  • Warranty3 year
3 Year WarrantyIP54 Rated

Key Features

  • Customize the four measurement channels with five different parameter modules
  • Large color graphic LCD with intuitive multilanguage interface
  • Non-volatle memory holds up to 2000 data points with time and date stamp
  • Easy-to-operate USB and RS232 ports and data transfer software included
  • Up to 5 point calibration with automatic recognition for USA/NIST and DIN buffers
  • Editing calibration errors with a one point recalibration
  • Selectable cell constant for 2- or 4-cell conductivity probes
  • Selectable reading reference temperature of 5, 10, 15, 20 or 25°C
  • Choice of practical salinity or natural sea water curves for salinity
  • Accepts both polarographic and RDO(optical/luminessence) probes
  • Automatic temperature and barometic pressure compensation, manual salinity compensation
  • 3 year warranty

More About this Item

The VERSA STAR advanced meter provides the versatility to operate from one to four measurement channels at the same time. The five parameter modules available for the channels including; pH, pH/ISE, Conductivity, DO/RDO, and pH with LogR. Customize the meter you need with the options of meter only and modules, a meter with module package, or a full kit with meter, modules, electrode and solutions.

This versatile meter has a large color graphic LCD display. The informative display shows readings with temperature, connections, time, date, sample ID, user ID and calibration data. Plain language screen prompts, soft keys, and a multilanguage interface supports English, Spanish, French, Italian, German, and Chinese.

Non-volatle memory holds up to 2000 data sets with time and date stamp. Easy-to-operate USB and RS232 ports and data transfer software take stress out of data management.

Adds two pH/ISE/Temperature, and one Conductivity/TDS/Salinity/Resistivity/Temperature, and one DO/Temperature measurement modules. Probes sold separately.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat