Thermo Scientific TSX3020EA Enzyme Freezer, 30 cu ft; 120V/60 Hz

Representative image only.
Thermo Scientific Mfr # TSX3020EA – Item # WW-44202-48
Store enzymes securely and sustainably in a –20°C manual-defrost freezer
  • ENERGY STAR® certified to save money and the environment
  • V-drive technology maintains temperature uniformity by detecting usage patterns
  • Whisper quiet with a sound level of 52 dBA
  • Easy mobility with four 2"casters; front two are lockable
  • Enzyme bins included for convenient storage
  • Capacity is 30 cubic feet
  • Operates on 120 VAC, 60 Hz power
  • 2 Years Parts & Labor Warranty

SPECS & DESCRIPTION

Specifications & Description

  • Freezer Capacity (cu ft)30
  • Freezer Capacity (Liters)827
  • Min Temperature - Freezer (° F)-13
  • Max Temperature - Freezer (° F)5
  • Min Temperature - Freezer (° C)-25
  • Max Temperature - Freezer (° C)-15
  • Temperature ControlMicroprocessor
  • Freezer Interior Width (in)30
  • Freezer Interior Height (in)58
  • Freezer Interior Depth (in)28 1/2
  • Freezer Interior Width (cm)76
  • Freezer Interior Height (cm)147
  • Freezer Interior Depth (cm)72
  • Exterior Width (in)34
  • Exterior Height (in)7 1/2
  • Exterior Depth (in)38 7/8
  • Exterior Width (cm)86
  • Exterior Height (cm)199
  • Exterior Depth (cm)99
  • Number of Doors1
  • Number of Shelves9
  • Number of Boxes54
  • AlarmYes
  • Power (VAC)115
  • Power (Hz)60
  • DescriptionEnzyme Freezer, 30 cu ft; 120V/60 Hz
  • Warranty2 years parts & labor
2 Year WarrantyUL Listed to Canadian safety standards.Made in the U.S.A.

Includes

Enzyme bins, USB drive with user manuals, certificate of conformance, control panel keys, cabinet door keys, and power cord.

Key Features

  • ENERGY STAR® certified to save money and the environment
  • V-drive technology maintains temperature uniformity by detecting usage patterns
  • Whisper quiet with a sound level of 52 dBA
  • Easy mobility with four 2"casters; front two are lockable
  • Enzyme bins included for convenient storage
  • Capacity is 30 cubic feet
  • Operates on 120 VAC, 60 Hz power
  • 2 Years Parts & Labor Warranty

More About this Item

Safely store temperature sensitive enzymes without the risk of damaging temperature fluctuations associated with auto defrost units.

Designed to support sample protection and sustainability objectives, TSX Series high-performance -20°C manual defrost enzyme freezers feature unique V-drive technology. This facilitates ultimate temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising sample integrity.

Features include easy-to-use microprocessor control with setpoint security and alarms, ergonomic door handle and compatibility with SMART VUE® wireless monitoring systems.

In addition to being ENERGY STAR® certified, natural R290 refrigerants make the TSX Series compliant with the U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP).

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat