Thermo Scientific TSG General Purpose Refrigerator, 49 cuft, Gray Exterior, Double Glass Doors; 115 VAC

Representative image only.
Thermo Scientific Mfr # TSG49RSGA – Item # WW-44261-27
Go green while improving cold storage
  • EPA ENERGY STAR® certified high-efficiency cooling system
  • Green, natural refrigerants offer greater safety
  • Manufacured in a zero-waste-to-landfill facility
  • Whisper-quiet compressor and low heat output fits in any lab setting
  • Temperature high and low, door ajar, and power failure alarms protect critical supplies and samples
  • Capacity is 49 cubic feet Exterior is gray Glass double doors

SPECS & DESCRIPTION

Specifications & Description

  • Refrigerator Capacity (cu ft)49
  • Refrigerator Capacity (Liters)1387
  • Min Temperature - Refrigerator (° C)2
  • Max Temperature - Refrigerator (° C)10
  • Temperature ControlDigital
  • Refrigerator Interior Width (in)49
  • Refrigerator Interior Height (in)61 3/4
  • Refrigerator Interior Depth (in)28 1/2
  • Refrigerator Interior Width (cm)124.5
  • Refrigerator Interior Height (cm)156.9
  • Refrigerator Interior Depth (cm)72.4
  • Exterior Width (in)27 3/4
  • Exterior Height (in)81
  • Exterior Depth (in)32
  • Exterior Width (cm)70.5
  • Exterior Height (cm)205.7
  • Exterior Depth (cm)81.3
  • Number of Doors2 glass
  • Number of Shelves8
  • AlarmYes
  • Cabinet ConstructionSteel
  • Power (VAC)115
  • Power (Hz)60
  • Power (amps)5
  • Power (Breaker)15 A
  • DescriptionGeneral Purpose Refrigerator, 49 cuft, Gray Exterior, Double Glass Doors; 115 VAC
UL Listed to Canadian safety standards.Made in the U.S.A.UL Listed.

Key Features

  • EPA ENERGY STAR® certified high-efficiency cooling system
  • Green, natural refrigerants offer greater safety
  • Manufacured in a zero-waste-to-landfill facility
  • Whisper-quiet compressor and low heat output fits in any lab setting
  • Temperature high and low, door ajar, and power failure alarms protect critical supplies and samples
  • Capacity is 49 cubic feet Exterior is gray Glass double doors

More About this Item

Ideal for storing reagents, media, and samples, these general purpose refrigerators are also engineered to be better for the environment. Save electricity and money with adaptive defrost, decreased compressor cycle time, and reduced heat transfer. The natural refrigerant R290 will not deplete the ozone layer or contribute substantially to climate change. Being manufactured in a zero-waste-to-landfill facility ensures at least 99% of the waste produced is reused, recycled, composted, incinerated, or used in energy recovery.

The refrigerator's quiet 51-dBA operation and low heat output allows for installation near workstations for added convenience. Visible and audible alarms when temperature exceeds high or low limits, when the doors are ajar, and when there is a power outage prevent losses of valuable contents. Durable steel construction withstands chemical spills and resists corrosion.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat