Thermo Scientific REL-2304D Laboratory upright refrigerator (single door); 23.3 CU FT; 200/230V
Representative image only.
Thermo Scientific Mfr # REL2304D – Item # WW-44185-25
UL and cUL listed, TUV approved, and CE marked
  • 13 month warranty
Stay Connected - Monitor Your Refrigerator or Freezer Temperatures
SPECS & DESCRIPTION
Specifications & Description
  • Freezer Capacity (cu ft)20.2
  • Freezer Capacity (Liters)572
  • Min Temperature - Freezer (° F)-123
  • Max Temperature - Freezer (° F)-58
  • Min Temperature - Freezer (° C)-86
  • Max Temperature - Freezer (° C)-50
  • Temperature ControlMicroprocessor
  • Number of Doors1
  • Number of Shelves5
  • AlarmYes
  • DisplayLED
  • Power (VAC)115
  • Power (Hz)60
  • Power (amps)16
  • Power (Breaker)20 A
  • DescriptionLaboratory upright refrigerator (single door); 23.3 CU FT; 200/230V
  • Warranty13 month
Conformite EuropeenneCanadian Standards AssociationUL Listed.
Key Features
  • 13 month warranty
More About this Item
Store samples for industrial or scientific applications with this Thermo Scientific™ Revco™ High-Performance Laboratory Refrigerator. Fridge features a microprocessor control and positive airflow systems, industrial-quality cabinet construction, and heavy-duty refrigeration compressors.