Thermo Scientific PC 300 Immersion Circulator, thermostat with bridge (230VAC/50Hz)

Representative image only.
Thermo Scientific Mfr # 1590025 – Item # WW-12135-57
Wide range of performance levels for up to 50-liter baths
  • Easy to use and energy efficient—rotate control head 90° for easier viewing
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Max Temperature (° C)300
  • Min Temperature (° C)Ambient
  • Pump TypeForce and suction
  • Max Flow Rate (L/min)20
  • Low-Level CutoffYes
  • High-Temperature CutoffYes
  • Immersion Depth (in)5 3/4
  • Temperature ControlPID
  • Temperature SensorPt RTD
  • Pressure Pump Max Head At 0 Flow (PSI)6.9
  • Suction PumpYes
  • Wetted MaterialsEPDM, PEI, PPS, SS, Vectra® and Viton®
  • Power (VAC)230
  • Power (Hz)50
  • DescriptionImmersion Circulator, thermostat with bridge (230VAC/50Hz)
  • Warranty3 year
3 Year WarrantyConformite Europeenne

Key Features

  • Easy to use and energy efficient—rotate control head 90° for easier viewing
  • 3 year warranty

More About this Item

Advanced (AC) Controllers have all the same features as the SC models, but also include a powerful three-speed force/suction pump for external loop applications. You get an additional five languages (Spanish, Italian, Chinese, Russian, and Japanese). They have an internal and external temperature control mode (remote sensor, NAMUR type). Features include a programmable application temperature alarm with user selected audible alarm and a “go-to-safe-state” or shut off option. The AC 200 models offer ramp programming with edit during ramp feature and a timer with real-time clock for time-critical applications. You can customize the control parameters for external and internal temperature control via serial communication (the PID-portions can be changed this way). There is also an option for extra communication for Profibus and CAN bus analog I/O—call for information.

Premium (PC) Controllers encompass all of the features of the SC and AC baths, and then provide greater control and flexibility. Pump speed adjusts to 10 different levels for turbulence control. Use the 10-key access keypad for fast and intuitive software navigation. User-level management allows you to lock in high and low temperature limitations for specific applications—prevents inadvertent changes and allows you to quickly recall saved settings. Control parameters are optimized for the application behavior, so that temperature control responds rapidly with zero over swing or under swing. For datalogging, all ring buffer data is stored for serial communication. Extra communication option for Profibus and CAN bus analog I/O—call for information.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat