Thermo Scientific Orion Star T930 Ion Titrator Chloride/Salt Kit; 100 to -240 VAC

Representative image only.
Thermo Scientific Mfr # START9301 – Item # WW-94102-36
Get fast, reliable, and reproducible automated titrations
  • Provides dedicated ion concentration titrations
  • MKA mode performs automated calibrations
  • Program and save up to 10 user-defined methods
  • Store up to 100 titration data sets with date/time stamp
  • Easily transfer data to printer, computer, or USB drive
  • Minimize downtime with replaceable burettes, tubing, and dispensers
  • Large easy-to-ready touchscreen display enables fast setup
  • Perform accurate measurements with silver billet electrode
  • 1 year warranty
Often bought with...

SPECS & DESCRIPTION

Specifications & Description

  • UnitsmV, ISE
  • Display TypeTouchscreen
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1
  • mV Accuracy±0.2
  • Min Temperature (° C)-5
  • Max Temperature (DegreeC)100
  • Temperature Resolution0.1°F (0.1°C)
  • Temperature Accuracy±0.2°C
  • Capacity (mL)20
  • OutputUSB
  • Data LoggingYes
  • Memory Size100 data sets
  • Power (VAC)110 to 240
  • Power (Hz)50/60
  • Length (in)10
  • Width (in)16
  • Height (in)14
  • Length (cm)25.4
  • Width (cm)40.6
  • Height (cm)35.6
  • DescriptionIon Titrator Chloride/Salt Kit; 100 to -240 VAC
  • Warranty1 Year
1 Year WarrantyConformite Europeenne

Includes

Silver billet electrode, electrode cable, 20 mL burette, stirrer probe, dispenser probe, standard tubing kit, 1 L plastic bottle, GL38 bottle cap with desiccant tube, computer cable, literature on USB drive, and 110 to 240 V power supply.

Key Features

  • Provides dedicated ion concentration titrations
  • MKA mode performs automated calibrations
  • Program and save up to 10 user-defined methods
  • Store up to 100 titration data sets with date/time stamp
  • Easily transfer data to printer, computer, or USB drive
  • Minimize downtime with replaceable burettes, tubing, and dispensers
  • Large easy-to-ready touchscreen display enables fast setup
  • Perform accurate measurements with silver billet electrode
  • 1 year warranty

More About this Item

The T900 series automated potentiometric titrators feature a state-of-the-art reagent dispensing system designed to simplify your titrations and offer more reliable, reproducible, and faster results then manual titrations. These compact, rugged titrators are simple to use, let you easily navigate live titration analysis, and are ideal for small bench spaces. You can safely run the titrator unattended until titration is completed.

The automated titrant delivery system, featuring a high-accuracy burette and dispenser, provides consistent results every time. Internal data storage maintains titration history, titrant standardizations, electrode calibrations, and direct measurements, all of which can easily be transferred to an external device.

T930 ion titrators offer dedicated ion concentration titrations including salt in food products, chloride in drinking water and wastewater, ammonia and Total Kjeldajhl Nitrogen (TKN) of wastewater, surfactants in consumer products, and total hardness of drinking water and wastewater. Titration techniques include equivalence point titrations, preset mV endpoint titrations, and multiple known addition (MKA) mode.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat