Thermo Scientific AQ3010 Turbidity meter, complete kit
Representative image only.
Thermo Scientific Mfr # AQ3010 – Item # WW-99570-15
Faster calibration and measurements with user-friendly interface
  • Compliant with ISO 7027 IR LED, ISO 7027, and DIN 27027 standards
  • Nephelometric measurement with autoranging capabilities
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypePortable
  • Display typeLED
  • Min Turbidity0.01 to 1000
  • Turbidity Resolution0.01, 0.1, or 1 NTU
  • WaterproofYes
  • Battery4 x AAA batteries (included)
  • DescriptionTurbidity meter, complete kit
  • Warranty3 year
3 Year WarrantyConformite EuropeenneIP 67
Key Features
  • Compliant with ISO 7027 IR LED, ISO 7027, and DIN 27027 standards
  • Nephelometric measurement with autoranging capabilities
  • 3 year warranty
More About this Item
The Thermo Scientific AQUAfast AQ3010 turbidity meter is ideal for both lab and field measurements. A user-friendly interface, large LCD, and 0 to 1000 NTU automatic ranging make it easy to determine the clarity of your sample.
Calibration is simple with step-by-step prompts. This handheld meter is supplied with ready-to-use EPA-approved standards for calibration and comes with everything you need. The field case provides convenient storage and transport for the waterproof meter and accessories.