Thermo Scientific Nunc 376589 CryoTube Vial Holder 03755-71
Representative image only.
Thermo Scientific Nunc Mfr # 376589 – Item # WW-03755-71
₹13,300.00INR / Pkg of 1
SPECS & DESCRIPTION
Specifications & Description
  • Tubes Held40
  • Tube Size1.0 to 5.0 mL
  • ColorBlue
  • Height (in)7/8
  • Height (cm)2.2
  • DescriptionCryoTube Vial Holder 03755-71
More About this Item
This holder features an interlocking design for one-handed operation when using with CryoTube® starfooted. Holder is constructed of polyphenylene oxide and accepts up to forty 1.0- to 5.0-mL tubes. Measures 7 7/8"L x 4"W x 7/8"H. Autoclavable.