Thermo Scientific Nunc Disposable Inoculating Needle, Polystyrene, Sterile, Yellow; 600/Pk
Representative image only.
Thermo Scientific Nunc Mfr # 254399 – Item # WW-14213-21
Color-coded for quick and easy identification
  • Radiation sterilized—no need for flame sterilization
  • Hydrophilic treatment during manufacturing process
  • Liquid adherence properties that prevent accidental discharge of droplets
PRODUCT OPTIONS
₹47,700.00INR / Pkg of 600
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Handle TypeNeedle
  • ColorYellow
  • DescriptionDisposable Inoculating Needle, Polystyrene, Sterile, Yellow; 600/Pk
Key Features
  • Radiation sterilized—no need for flame sterilization
  • Hydrophilic treatment during manufacturing process
  • Liquid adherence properties that prevent accidental discharge of droplets
More About this Item
These sterile, polystyrene loops and needles are calibrated using a standard urea solution. A Thermo Scientific™ Nunc™ calibration certificate can be obtained upon request. With no need to flame, these loops and needles save you time. The risk of contamination is eliminated since these instruments are disposable. The surface of each loop and needle receives a special hydrophilic treatment to enhance the adhesion of droplets and prevent accidental discharge. Loops are color coded for easy identification.