Thermo Scientific Nunc Cell Culture Flask, Polystyrene, Sterile, Treated, Straight Neck, Filter Cap, 175 cm2; 32/Cs
Representative image only.
Thermo Scientific Nunc Mfr # 178883 – Item # WW-01932-44
Nunclon™ Delta surface treatment provides optimal cell attachment and growth
  • Virgin polystyrene provides excellent optical quality
  • Sterilized by gamma radiation
  • Large-bore necks—easy pipette and cell scraper access
PRODUCT OPTIONS
₹76,900.00INR / Case of 32
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolystyrene
  • Cap TypeFilter
  • Cap MaterialHDPE
  • Neck TypeStraight
  • Working Volume (mL)68
  • Culture Area (cm²)175
  • Pore Size (µm)0.22
  • DescriptionCulture Flask, Polystyrene, Sterile, Treated, Straight Neck, Filter Cap, 175 cm2; 32/Cs
Nonpyrogenic
Key Features
  • Virgin polystyrene provides excellent optical quality
  • Sterilized by gamma radiation
  • Large-bore necks—easy pipette and cell scraper access
More About this Item
Achieve maximum adhesion for a broad range of cell types with Thermo Scientific™ Nunc™ Cell Culture Treated Flasks. These flasks receive the proprietary Nunclon™ Delta surface treatment to ensure consistent cell growth.
Vented caps promote sufficient gas exchange while filtered caps allow constant airflow and minimize chances of contamination.
The unique hydrophobic 0.22µm filter cap utilizes a three-dimensional matrix of nonwettable polyethylene fibers that prevent penetration of microorganisms.