Thermo Scientific Nunc Cell Culture Flask, Polystyrene, Sterile, Treated, Straight Neck, Vent Cap, 80 cm2; 50/Cs
Representative image only.
Thermo Scientific Nunc Mfr # 153732 – Item # WW-01930-42
Nunclon™ Delta surface treatment provides optimal cell attachment and growth
  • Virgin polystyrene provides excellent optical quality
  • Sterilized by gamma radiation
  • Large-bore necks—easy pipette and cell scraper access
PRODUCT OPTIONS
₹61,400.00INR / Case of 50
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolystyrene
  • Cap TypeVented
  • Cap MaterialHDPE
  • Neck TypeStraight
  • Working Volume (mL)30
  • Culture Area (cm²)80
  • DescriptionCulture Flask, Polystyrene, Sterile, Treated, Straight Neck, Vent Cap, 80 cm2; 50/Cs
Nonpyrogenic
Key Features
  • Virgin polystyrene provides excellent optical quality
  • Sterilized by gamma radiation
  • Large-bore necks—easy pipette and cell scraper access
More About this Item
Achieve maximum adhesion for a broad range of cell types with Thermo Scientific™ Nunc™ Cell Culture Treated Flasks. These flasks receive the proprietary Nunclon™ Delta surface treatment to ensure consistent cell growth.
Vented caps promote sufficient gas exchange while filtered caps allow constant airflow and minimize chances of contamination.