Thermo Scientific Nunc 132867 TripleFlasks, Nunclon, 500cm2, Straight neck,Vented Cap; 32/Cs
Representative image only.
Thermo Scientific Nunc Mfr # 132867 – Item # WW-01931-00
Get three flasks in one!
  • Three parallel growth Nunclon™ Delta surfaces provide a total culture area of 500 cm2!
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolystyrene
  • Cap TypeVented
  • Neck TypeStraight
  • Total Volume (mL)700
  • Culture Area (cm²)500
  • DescriptionTripleFlasks, Nunclon, 500cm2, Straight neck,Vented Cap; 32/Cs
Key Features
  • Three parallel growth Nunclon™ Delta surfaces provide a total culture area of 500 cm2!
More About this Item
These unique flasks offer three times the yield of standard 175 cm2 flasks in the same space, making them ideal for intermediate scale-up production. Harvest cells using protein digesting enzymes such as trypsin, collagenase, and pronase. The design of this flask prevents mechanical scraping. Total culture area is achieved by way of the three parallel layers and a straight neck design that works effectively within automation systems. TripleFlask receives the Nunclon™ Delta treatment, a proprietary cell culture surface treatment that offers maximum adhesion for a broad range of cell types.