Thermo Scientific Nalgene Wraparound Face Shield, Polycarbonate
Representative image only.
Thermo Scientific Nalgene Mfr # 6355-0001 – Item # WW-06767-00
Full-length, wrap-around shield sits away from face for easy breathing
  • Meets ANSI standard Z87.1
  • 1 year warranty
₹49,300.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorGray
  • Lens ColorClear
  • Lens CoatingUV protection
  • ComplianceANSI Z87.1; 29 CFR part 1910.1030
  • Width (in)7 51/64
  • Depth (in)11 39/64
  • DescriptionWraparound Face Shield, Polycarbonate
  • Warranty1 year
1 Year Warranty
Key Features
  • Meets ANSI standard Z87.1
  • 1 year warranty
More About this Item
Optically clear window of 0.06" thick polycarbonate meets ANSI standard Z87.1—protects against UV radiation from 200 to 360 nm. Adjustable headband. Compliance: ANSI Z87.1, 29 CFR Part 1910.1030