Thermo Scientific Nalgene Bio Tank Sanitary Mixer Mount
Representative image only.
Thermo Scientific Mfr # 2651-0200 – Item # WW-50301-70
  • 6″ (15.2cm) polypropylene screw closure with 2″ (5.1cm) sanitary ferrule welded in the center
  • 2″ (5.1cm) silicone gasket and PDVF True Union clamp
  • Autoclavable
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionNalgene Bio Tank Sanitary Mixer Mount
Key Features
  • 6″ (15.2cm) polypropylene screw closure with 2″ (5.1cm) sanitary ferrule welded in the center
  • 2″ (5.1cm) silicone gasket and PDVF True Union clamp
  • Autoclavable
More About this Item
Enable overhead mixing in a closed system with this unique, sanitary flange assembly. Thermo Scientific™ Nalgene™ Closed-Dome Bio Tank Closure with Mixer Support Assembly is for use with all sizes of Nalgene™ Closed-Dome Bio Tanks. Nalgene™ Biotech Mixer is not included.
Compatible with Thermo Scientific Nunc Cylindrical Polypropylene Closed-Dome Tanks, models 50301-52, -53, -54, and -56.