Thermo Scientific Nalgene Pipette Washer for Up To 24" L Pipets
Representative image only.
Thermo Scientific Nalgene Mfr # 5245-0050 – Item # WW-06190-00
Offers wide range of cycling speeds, operating with water input from 1.5 to 12 liters per minute
  • Water inlet at the top eliminates back-siphoning to prevent contamination
  • Durable HDPE will provide years of use in your lab
  • 1 year warranty
PRODUCT OPTIONS
₹129,000.00INR / Each
Upgrade to Complete System…
SPECS & DESCRIPTION
Specifications & Description
  • Height (in)38
  • Wetted MatterialHDPE and PVC
  • Compatible withPipets up to 24\" L
  • DescriptionPipette Washer for Up To 24\" L Pipets
1 Year Warranty
Key Features
  • Water inlet at the top eliminates back-siphoning to prevent contamination
  • Durable HDPE will provide years of use in your lab
  • 1 year warranty
More About this Item
HDPE washers include top inlet to prevent back-siphoning of contaminated water. Connect directly to drain line. Use water flow as low as 1.5 liters/minute.
Washers include 18” L x 1” ID inlet and 4’ x ½” ID PVC tubing.