Thermo Scientific Nalgene Pipette Cleaning Jars for Up To 24" L Pipettes
Representative image only.
Thermo Scientific Nalgene Mfr # 5242-0050 – Item # WW-06176-00
Soak or sterilize glass pipettes
  • Jars can be used with sulfuric acid/dichromate solution if acid concentration is 60% or less
  • Recommended cleaning solution formula shown on the jar's label is effective and lasts longer than standard solutions
  • Use independently or with 06155-00 basket
  • 1 year warranty
PRODUCT OPTIONS
₹67,100.00INR / Each
Upgrade to Complete System…
SPECS & DESCRIPTION
Specifications & Description
  • Height (in)27
  • Wetted MatterialHDPE
  • Compatible withPipettes up to 24\" L
  • DescriptionPipette Cleaning Jars for Up To 24\" L Pipettes
1 Year Warranty
Key Features
  • Jars can be used with sulfuric acid/dichromate solution if acid concentration is 60% or less
  • Recommended cleaning solution formula shown on the jar's label is effective and lasts longer than standard solutions
  • Use independently or with 06155-00 basket
  • 1 year warranty
More About this Item
Ideal for soaking and washing pipettes, these cross-linked high-density polyethylene jars have a flanged base for good stability. Use independently or use with baskets and washers. Covers (included) convert jars into handy storage containers.