Thermo Scientific Nalgene 5040-0002 Cryoware labels; 1" x 2", 200/pack
Representative image only.
Thermo Scientific Nalgene Mfr # 5040-0002 – Item # WW-08282-31
Ensure accurate record keeping of cryogenic vials and storage boxes
  • Cloth labels adhere to plastics and cardboard
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCryoware labels; 1\" x 2\", 200/pack
  • Warranty1 year
1 Year Warranty
Key Features
  • Cloth labels adhere to plastics and cardboard
  • 1 year warranty
More About this Item
Once applied, labels will not peel or shrink, even under cryogenic conditions. Each resealable package contains 10 sheets of plain labels; 20 labels per sheet. Label size is 1" x 2". Pack of 200.