Thermo Scientific Nalgene Filling/Venting Cap for 1/4" Tubing, 53B
Representative image only.
Thermo Scientific Nalgene Mfr # 2162-0531 – Item # WW-06258-10
Transfer aseptic liquid safely
  • Autoclavable
PRODUCT OPTIONS
₹12,400.00INR / Each
Often bought with…
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene(PP)
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper Size53B
  • Cap ColorWhite
  • AutoclavableYes
  • Tubing ID6.4 mm
  • Descriptionfilling/venting cap for 1/4\" tubing, 53B
Key Features
  • Autoclavable
More About this Item
Filling/venting caps have thermoplastic elastomer (TPE) gaskets to allow aseptic liquid transfer.
Include 6" (15 cm) and 30" (76 cm) lengths of platinum-cured silicone tubing.