Thermo Scientific Hypersil 31603-104030 ODS-2 (C18) Column - Length 100mm, 4.0mm ID, 3µm particle size
Representative image only.
Thermo Scientific Hypersil Mfr # 31603-104030 – Item # WW-01890-17
High-efficiency columns with proven results and reproducibility
  • Suitable for Octadecyl silane (C18) phase
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhaseOctadecyl silane (C18)
  • Column Particle Size (µm)3
  • Tubing Inside Diameter (ID) (mm)4
  • DescriptionODS-2 (C18) Column - Length 100mm, 4.0mm ID, 3µm particle size
  • Warranty1 month
1 Month WarrantyISO9001 Certified Supplier
Key Features
  • Suitable for Octadecyl silane (C18) phase
  • 1 month warranty
More About this Item
Thermo Scientific™ Hypersil™ columns have been trusted by researchers for over 30 years. Each batch of silica produced for these columns is tested to make sure it results in the same separations as previous batches, guaranteeing you have the same results time after time, even after changing your column. The wide range of phases ensures a column is available to match your application.
Hypersil ODS-2 C18 Columns are manufactured under strict ISO 9001:2000 guidelines and provide a dependable L1 alternative. These columns are tested to confirm reproducibility and ruggedness required in QA and QC laboratories. Provide separations over a wide range of applications including polar compounds, vitamins, and amino acids. Stable in pH range of 2 to 8.