Thermo Scientific Hypersil 30905-104030 Phenyl Column - Length 100mm, 4.0mm ID, 5µm particle size
Representative image only.
Thermo Scientific Hypersil Mfr # 30905-104030 – Item # WW-01889-68
High-efficiency columns with proven results and reproducibility
  • Suitable for Phenyl phase
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhasePhenyl
  • Column Particle Size (µm)5
  • Tubing Inside Diameter (ID) (mm)4
  • DescriptionPhenyl Column - Length 100mm, 4.0mm ID, 5µm particle size
  • Warranty1 month
1 Month WarrantyISO9001 Certified Supplier
Key Features
  • Suitable for Phenyl phase
  • 1 month warranty
More About this Item
Thermo Scientific™ Hypersil™ columns have been trusted by researchers for over 30 years. Each batch of silica produced for these columns is tested to make sure it results in the same separations as previous batches, guaranteeing you have the same results time after time, even after changing your column. The wide range of phases ensures a column is available to match your application.
Hypersil Phenyl and Phenyl-2 Columns have retention characteristics similar to the MOS columns and are recommended for the separation of certain aromatic compounds and moderately polar compounds. Hypersil Phenyl-2 is endcapped. Stable in pH range of 2 to 8.