Thermo Scientific Hypersil 25402-202130 GOLD PFP Column - Length 200mm, 3.0mm ID, 1.9µm particle size
Representative image only.
Thermo Scientific Hypersil Mfr # 25402-202130 – Item # WW-01882-25
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhasePerfluorinated phenyl
  • Column Particle Size (µm)1.9
  • Tubing Inside Diameter (ID) (mm)2.1
  • DescriptionGOLD PFP Column - Length 200mm, 3.0mm ID, 1.9µm particle size
  • Warranty1 month
1 Month Warranty
Key Features
  • 1 month warranty
More About this Item
Thermo Scientific Hypersil GOLD PFP Columns provide excellent peak shape and selectivity of fluorinated and other halogenated compounds. These columns are also particularly useful for non-halogenated aromatic compounds containing hydroxyl, carboxyl, nitro, or other polar groups; or in separating difficult mixtures containing closely related halogenated and polar compounds. Stable in a pH range of 2 to 8.