Thermo Scientific Hypersil 25302-202130 GOLD aQ Column - Length 200mm, 2.1mm ID, 1.9µm particle size
Representative image only.
Thermo Scientific Hypersil Mfr # 25302-202130 – Item # WW-01880-30
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhaseOctadecyl silane (C18), aqueous
  • Column Particle Size (µm)1.9
  • Tubing Inside Diameter (ID) (mm)2.1
  • DescriptionGOLD aQ Column - Length 200mm, 2.1mm ID, 1.9µm particle size
  • Warranty1 month
1 Month Warranty
Key Features
  • 1 month warranty
More About this Item
Thermo Scientific Hypersil GOLD aQ Columns offer a polar endcapped C18 phase that provides the same optimized peak shape and resolution with the added selectivity and retention of polar compounds. Ideal for the quantitative analysis of polar analytes present in trace amounts and for reversed phase applications using 100% aqueous mobile phase. Stable in a pH range of 2 to 9.