Thermo Scientific DNA Surface Decontaminant; 250 mL Bottle
Representative image only.
Thermo Scientific Mfr # 7010 – Item # WW-25715-04
Fast, effective way to remove RNase and DNA contamination
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDNA surface decontaminant, 250 ml bottle
More About this Item
These surface decontaminants provide a safe, fast, and proven alternative to remove nuclease and DNA contamination from benchtops, instruments, pipettors, glass, and plasticware. Conventional methods to remove RNA and DNA contamination use a Diethylpyro-carbonate (DEPC) treatment, which is suspected to be carcinogenic and must be handled with gloves and under approved fume
hoods at all time.
DNA AWAY requires no prep time and is ready to use straight out of the 250-mL bottle.