Thermo Scientific CY50945 8 Cryobiological Inventory Systems
Representative image only.
Thermo Scientific Mfr # CY50945 – Item # WW-05094-50
Store samples up to 185 days without replenishing LN2
  • Compact size stores up to 3600 samples
  • Advanced vacuum insulation minimizes liquid nitrogen evaporation
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Volume (Liters)111
  • Outside Diameter (OD) (in)22
  • Outside Diameter (OD) (cm)55.9
  • Static Holding Time (days)185
  • Evaporation (Liters/day)0.6
  • Tubes Held1800
  • Height (in)37 1/2
  • Height (cm)95.2
  • Description8 Cryobiological Inventory Systems
  • Warranty1 year
1 Year Warranty
Key Features
  • Compact size stores up to 3600 samples
  • Advanced vacuum insulation minimizes liquid nitrogen evaporation
  • 1 year warranty
More About this Item
Find the sample you need quickly and efficiently! All Thermo Scientific Locator® systems feature a double-walled, vacuum insulated vessel with a narrow-mouth neck that keeps liquid nitrogen losses to a minimum. Stainless steel racks fit snugly against the wall of the Thermo Scientific Locator® system to hold multiple racks. The rack handles rest in numbered slots for easy identification and retrieval. Each rack has individual storage locations for cryogenic boxes.
Use Thermo Scientific Locator® Jr. system (model 05094-60) to fit in tight spaces - this scaled-down version is only 26" in height! Thermo Scientific Locator® Jr. system holds four racks; each rack holds four 5-1/4" square cryogenic boxes. Select Locator 4 system (model 05094-40) to hold up to four racks; each rack holds nine 51⁄4" (13.3 cm) square cryogenic boxes. The Locator 8 system (model 05094-50) holds up to eight racks; each rack holds nine 3" (7.6 cm) square cryogenic boxes.
Order optional roller cart to easily transport your Locator system.
The Locator alarm alerts you to low liquid nitrogen level.