Thermo Scientific BioBasic SEC 1000 Guard Column - 30mm by 7.8mm, 1000 Angstrom pore size

Representative image only.
Thermo Scientific BioBasic Mfr # 73605-037821 – Item # WW-01902-16
Provide superior separation of water soluble compounds
  • Ideally suited for sample clean-up
  • Analyte separation over a wide range of molecular weights
  • Long, efficient column life
  • 1 month warranty

SPECS & DESCRIPTION

Specifications & Description

  • PhaseSilica
  • Column Length (mm)30
  • Column Inside Diameter (ID) (mm)7.8
  • Column Particle Size (µm)0.1
  • DescriptionSEC 1000 Guard Column - 30mm by 7.8mm, 1000 Angstrom pore size
1 Month Warranty

Key Features

  • Ideally suited for sample clean-up
  • Analyte separation over a wide range of molecular weights
  • Long, efficient column life
  • 1 month warranty

More About this Item

BioBasic SEC (Size Exclusion Chromatography) columns are made with a mechanically rigid silica that does not swell or shrink with changes in solvent, providing higher efficiencies than polymer-based columns. The highly base-deactivated 5µm silica is polymer-coated to ensure separation only occurs on the basis of molecular size. Columns provide highly efficient separation of compounds with a molecular weight of 100 to 10,000,000.
Each lot of silica used in the production of BioBasic SEC columns is tested with a range of proteins to confirm accuracy. Each column is also tested for compliance with efficiency specifications and is shipped with a test certificate with both the column and silica efficiency data.
BioBasic SEC columns are ideal for gel filtration separation of proteins an

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat