Thermo Scientific BioBasic 72405-254030 Phenyl Column - Length 250mm, 4.0mm ID, 5µm particle size
Representative image only.
Thermo Scientific BioBasic Mfr # 72405-254030 – Item # WW-01899-26
Provide alternate selectivity, especially for peptides with aromatic side chains
  • Lower carbon loading for retention of larger proteins and peptides
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhasePhenyl
  • Column Particle Size (µm)5
  • Tubing Inside Diameter (ID) (mm)4
  • DescriptionPhenyl Column - Length 250mm, 4.0mm ID, 5µm particle size
  • Warranty1 month
1 Month Warranty
Key Features
  • Lower carbon loading for retention of larger proteins and peptides
  • 1 month warranty
More About this Item
Thermo Scientific BioBasic HPLC columns provide improved performance for biological samples including peptides, proteins, and other biomolecules. The high purity silica and tailored pore sizes provide superior performance in terms of improved resolution, efficiency, reproducibility, and column life.
BioBasic Phenyl Columns contain 300Å pore size silica for maximum performance when used with biomolecules. These columns are ideally suited for the separation of high-hydrophobicity peptides nominally greater than 5,000 Da, and those with aromatic side chains. BioBasic Phenyl columns are also useful where alkyl chain chemistries do not provide adequate enough selectivity. The highly pure, deactivated silica in these columns contain extra dense bonding to provid