Thermo Scientific BioBasic 8 Column - Length 100mm, 2.1mm ID, 5µm particle size

Representative image only.
Thermo Scientific BioBasic Mfr # 72205-102130 – Item # WW-01898-08
Provide excellent separation of a wide range of peptides
  • Excellent starting column for peptide and protein separations
  • 1 month warranty

SPECS & DESCRIPTION

Specifications & Description

  • PhaseOctylsilane (C8)
  • Column Length (cm)10
  • Column Length (mm)100
  • Column Inside Diameter (ID) (mm)2.1
  • Column Particle Size (µm)5
  • Tubing Inside Diameter (ID) (mm)2.1
  • Description8 Column - Length 100mm, 2.1mm ID, 5µm particle size
  • Warranty1 month
1 Month Warranty

Key Features

  • Excellent starting column for peptide and protein separations
  • 1 month warranty

More About this Item

Thermo Scientific BioBasic HPLC columns provide improved performance for biological samples including peptides, proteins, and other biomolecules. The high purity silica and tailored pore sizes provide superior performance in terms of improved resolution, efficiency, reproducibility, and column life.

BioBasic 8 Columns contain 300Å pore size silica, which is ideally suited for medium-hydrophobicity peptides nominally less than 20,000 Da. Providing less hydrophobic retention than BioBasic 18 columns, BioBasic 8 columns are great starting columns for protein separations and may provide changes in elution patterns. Columns provide a highly stable, reproducible surface with sharp peak shap

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat