Thermo Scientific Barnstead D3751 Remote Dispenser Filter
Representative image only.
Thermo Scientific Barnstead Mfr # D3751 – Item # WW-99254-50
Use with Nanopure®, Nanopure RO, Easypure® II, E-Pure™, and B-Pure™ systems
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible withQuad-style Nanopure
  • Max Flow Rate (LPM)2
  • DescriptionRemote Dispenser Filter
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Final filters remove particulates down to 0.2 µm to provide bacteria- free water. Constructed of hollow fiber material, filters do not contain any wetting agents or surfactants. Filters are radiation sterilized and individually sealed.
Autoclavable.