Thermo Scientific Barnstead B440704 Auto Collection System
Representative image only.
Thermo Scientific Barnstead Mfr # B440704 – Item # WW-99292-70
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAuto Collection System
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Automate your purification system with the optional automatic collection system. A water-level sensor in the reservoir turns the heater and water supply on and off to keep water storage levels between 38 and 45 liters.