Thermo Scientific Barnstead 50132391 LabTower RO Water Purification System; 40L/Hr, 100L

Representative image only.
Thermo Scientific Barnstead Mfr # 50132391 – Item # WW-99289-45
Specfically developed for functional and economical production of high purity water
  • Integrated feed water monitoring ensures water quality and extends cartridge service life
  • Easy-to-operate clear display tilts for optimal viewing
  • Reservoir fill level (%) automatically shown on display
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • ASTM Water GradeType III
  • Max Production (L/hour)40
  • Max Flow Rate (L/hour)40
  • Inlet PortR 3/4\"
  • Feed Water Requirements (Source)potable tap water, softened or hardness stabilized
  • Feed Water Requirements (Conductivity)<1500 µS/cm
  • Feed Water Requirements (pH range)4 to 11
  • Feed Water Requirements (Temperature)2-35⁰C
  • Type III Water (Bacteria)99%
  • Min Operating Pressure (PSI)29
  • Max Operating Pressure (PSI)87
  • Min Operating Pressure (bar)2
  • Max Operating Pressure (bar)6
  • DescriptionLabTower RO Water Purification System; 40L/Hr, 100L
  • Warranty1 year
1 Year Warranty

Key Features

  • Integrated feed water monitoring ensures water quality and extends cartridge service life
  • Easy-to-operate clear display tilts for optimal viewing
  • Reservoir fill level (%) automatically shown on display
  • 1 year warranty

More About this Item

A complete mobile solution! This stand-alone unit with integrated 100-L reservoir requires no bench space and the smooth-rolling casters allow easy relocation in your lab.

Microprocessor control offers automatic operation, as well as real-time clock records system of errors and faults. The code-protected operating system prevents unauthorized changes to settings.

Use the RS-232 interface for safe documentation of all measured values and faults with date and time in compliance with GLP guidelines; order external printer separately.

Two production capacities easily handle your water requirements for reagent preparation, media preparation, and glassware rinsing. Dispense two ways - from reservoir or directly from system. Plus, an adjustable setting for reservoir volume allows decreased storage when demand is low. Purified water in the reservoir is continuously circulated to preserve water quality during activity.


Pretreatment packages, sterile overflow, and sterile vent are required and should be ordered separately.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat