Thermo Scientific Barnstead 50132387 Pacific RO Water Purification System 12 L/hr

Representative image only.
Thermo Scientific Barnstead Mfr # 50132387 – Item # WW-99289-20
Specifically developed for functional and economical production of high purity water
  • Integrated feed water monitoring ensures water quality and extends cartridge service life
  • Easy-to-operate clear display tilts for optimal viewing
  • Reservoir fill level (%) automatically shown on display
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • ASTM Water GradeType III
  • Max Production (L/hour)12
  • Max Flow Rate (L/hour)12
  • Inlet PortR 3/4\"
  • Feed Water Requirements (Source)potable tap water, softened or hardness stabilized
  • Feed Water Requirements (Conductivity)<1500 µS/cm
  • Feed Water Requirements (pH range)4 to 11
  • Feed Water Requirements (Temperature)2-35⁰C
  • Type III Water (Bacteria)99%
  • Min Operating Pressure (PSI)29
  • Max Operating Pressure (PSI)87
  • Min Operating Pressure (bar)2
  • Max Operating Pressure (bar)6
  • Height (in)13
  • Depth (in)14.625
  • Width (in)23.75
  • Height (cm)33
  • Depth (cm)37.2
  • Width (cm)60.3
  • DescriptionPacific RO Water Purification System 12 L/hr
  • Warranty1 year
1 Year WarrantyConformite Europeenne

Key Features

  • Integrated feed water monitoring ensures water quality and extends cartridge service life
  • Easy-to-operate clear display tilts for optimal viewing
  • Reservoir fill level (%) automatically shown on display
  • 1 year warranty

More About this Item

The Barnstead Pacific RO system delivers up to 240L/day of Type III (RO) water from a tap feed water supply. These units are available in five product water capacities - ideal for applications such as preparing and diluting buffers or supplying lab equipment like autoclaves, water baths, glassware washers, and incubators. Space-saving design allows for system installation on benchtop or for wall mounting.

Microprocessor control offers automatic operation, as well as real-time clock records system of errors and faults. The code-protected operating system prevents unauthorized changes to settings. Use the RS-232 interface for safe documentation of all measured values and faults with date and time in compliance with GLP guidelines; order external printer separately.

Opaque polyethylene reservoirs (required; order separately) allow complete draining and efficient cleaning and disinfection. Reservoirs are available with an optional recirculation pump which protects high purity water from bacterial growth during standstill.




REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat