Thermo Scientific Barnstead 06.5032 30L Tank with pump/display (Reservoir with Recirculation Pump)
Representative image only.
Thermo Scientific Barnstead Mfr # 06.5032 – Item # WW-99299-07
  • Space-saving design permits benchtop installation or wall mounting
  • Reservoir fill level automatically shown on dipslay
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • ASTM Water GradeType II
  • Capacity30 L
  • Description30L Tank with pump/display (Reservoir with Recirculation Pump)
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Space-saving design permits benchtop installation or wall mounting
  • Reservoir fill level automatically shown on dipslay
  • 1 year warranty
More About this Item
Opaque polyethylene reservoirs (required; order separately) allow complete draining and efficient cleaning and disinfection.
A sterile overflow, a sterile filter, and a CO2 Absorber and sterile filter are required for complete system. Order separately.
Reservoirs with a recirculation pump protects high purity water from bacterial growth during standstill.
30L reservoirs can also be used with the optional wall mounting bracket for further space-saving capabilities.