Thermo Scientific AQUAFAST Total Nitrogen Tube, Low Level, 50 Tests
Representative image only.
Thermo Scientific Mfr # ACD004 – Item # WW-99570-12
  • Each kit includes 50 vials with pre-measured reagents
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Test ParametersTotal nitrogen
  • RangeLow level
  • DescriptionTotal Nitrogen Tube, Low Level, 50 Tests
  • Warranty1 year
1 Year Warranty
Key Features
  • Each kit includes 50 vials with pre-measured reagents
  • 1 year warranty
More About this Item
Kits require a reactor for digestion (99581-35), and a colorimeter to read results.