Thermo Scientific A3-1 Owl Recirculating Cell, 23 x 40 cm gel size

Representative image only.
Thermo Scientific Mfr # A3-1 – Item # WW-28553-40
Longer path length for more detailed DNA/RNA analysis
  • Base functions as a built-in three-point leveling platform for flat, uniformly thick gels
  • Pour and run gels right in the gasketed gel tray — no taping or sealing needed
  • UV transparent gel tray lets you view DNA bands after performing runs
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Buffer Solution Required (mL)4500
  • Gel Length (cm)40
  • Gel Thickness (mm)1.5
  • Gel Width (cm)23
  • Cell Width (in)11
  • Cell Height (in)4
  • Cell Depth (in)20.88
  • Cell Width (cm)27.94
  • Cell Height (cm)10.16
  • Cell Depth (cm)53.04
  • DescriptionOwl Recirculating Cell, 23 x 40 cm gel size
  • Warranty3 year
3 Year Warranty

Key Features

  • Base functions as a built-in three-point leveling platform for flat, uniformly thick gels
  • Pour and run gels right in the gasketed gel tray — no taping or sealing needed
  • UV transparent gel tray lets you view DNA bands after performing runs
  • 3 year warranty

More About this Item

Self-recirculating models feature barbed fitting ports for buffer exchange via an external recirculating pump to minimize buffer breakdown and maintain constant pH.


What's included: buffer chamber, UV transparent gel tray with gasket end gates, safety lid with attached power leads, and built-in leveling base. All combs are 1.5-mm thick.
Item 28553-40 also includes two 25-well and two 50-well combs.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat