Thermo Scientific 88880017 LP Vortex Mixer, 100-240 VAC; US Plug
Representative image only.
Thermo Scientific Mfr # 88882009 – Item # WW-04725-01
Low-profile design with small footprint easily stores away when not in use
  • Choose between continuous or pulse mode
  • Simple dial controls speed from 0 to 3000 rpm
  • Automatic overload shutoff
  • Sturdy design and weight keeps unit from "walking" while in use
  • Variety of accessories accommodates different-sized vessels
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Power (VAC)100 to 240
  • DescriptionLP Vortex Mixer, 100-240 VAC; US Plug
  • Warranty2 year
Two year warranty
Key Features
  • Choose between continuous or pulse mode
  • Simple dial controls speed from 0 to 3000 rpm
  • Automatic overload shutoff
  • Sturdy design and weight keeps unit from "walking" while in use
  • Variety of accessories accommodates different-sized vessels