Thermo Scientific 7000 Aquamate Vis Spectrometer

Representative image only.
Thermo Scientific Mfr # AQ7000 – Item # WW-99564-10
Designed for water and wastewater analysis
  • Over 260 pre-programmed water quality methods plus user customizable methods
  • Methods stored on USB stick to be added or removed as needed
  • One-point calibration adjustment ensures the highest accuracy
  • Compatible with several leading brands of reagents
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Source LampTungsten-Halogen
  • Source Lamp Life1000 hours
  • Beam TypeSingle
  • Bandwidth5.0 nm
  • Min Wavelength (nm)325
  • Max Wavelength (nm)1100
  • Wavelength Accuracy±1.0 nm
  • Min Photometric - Transmittance (%T)0
  • Max Photometric - Transmittance (%T)125
  • Min Photometric - Absorbance (A)0
  • Max Photometric - Absorbance (A)3
  • Min Photometric - Concentration (C)-9999
  • Max Photometric - Concentration (C)9999
  • DetectorSilicon photodiode
  • OutputUSB
  • Display TypeLCD
  • Width (cm)29.9
  • Height (cm)23.7
  • Width (in)11 3/4
  • Height (in)9 21/64
  • Power (Hz)50/60
  • DescriptionAquamate Vis Spectrometer
  • Warranty1 year
1 Year WarrantyConformite Europeenne

Key Features

  • Over 260 pre-programmed water quality methods plus user customizable methods
  • Methods stored on USB stick to be added or removed as needed
  • One-point calibration adjustment ensures the highest accuracy
  • Compatible with several leading brands of reagents
  • 1 year warranty

More About this Item

AquaMate spectrophotometers offer the accuracy, dependability and ease of use your lab requires. Compact design with intuitive operation targeted to water and wastewater analysis. The pre-programmed or user-customizable water quality methods offer absorbance, % transmittance and concentration testing. Ensure wavelength accuracy and quality measurements with performance verification testing in accordance to GLP and GMP.

Aquamate features three USB connectors, providing the option to store methods and data to a USB stick, transfer data to a computer, or print data to an external printer. A wide range of vial sizes can be used with the 3 position cell-holder (turret): 13 to 25 mm round vials, 20 to 50 mm rectangular vials, and a 10 mm square vial. System is compatible with Orion, Merck, Chemetrics, and Hach chemistries.



AQ7000 Vis Spectrophotometer is the cost-effectve solution for accurate visible measurements. It offers 325 to 1100 nm selectable wavelengths, ideal for most concentration measurements. Combines an intuitive user interface, tungsten-halogen lamp, and simple single-beam optical geometry for accurate, reproducible results.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat