Thermo Scientific 68700 SnakeSkin Pleated Dialysis Tubing, 7000 MWCO, 22mm ID; 35ft

Representative image only.
Thermo Scientific Mfr # 68700 – Item # WW-02904-05
Scale down your dialysis prep time for large sample volumes from 12 to 100 mL
  • The pleated format makes it easy to open and ready to use, streamlining dialysis preparation, save considerable time over dry, flat tubing.
  • Regenerated cellulose is ideal for desalting, buffer exchange and removal of small molecular weight contaminants.
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)7000
  • Diameter (mm)22
  • Length Per Pack (ft)35
  • Diameter (in)0.866
  • DescriptionSnakeSkin Pleated Dialysis Tubing, 7000 MWCO, 22mm ID; 35ft
  • Warranty1 year
1 Year Warranty

Key Features

  • The pleated format makes it easy to open and ready to use, streamlining dialysis preparation, save considerable time over dry, flat tubing.
  • Regenerated cellulose is ideal for desalting, buffer exchange and removal of small molecular weight contaminants.
  • 1 year warranty

More About this Item

The dialysis tubes are supplied in 8¡± sticks containing 35 feet of 22 mm internal diameter tubing. The amount of tubing required can be calculated using 3.7 mL sample per cm of dry tubing. To use it, a researcher simply pulls out the required length of tubing, cuts it off and applies a closure. The sample is then added through the other end of the dry tubing and the second closure is applied.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat