Thermo Scientific 5107BNMD No-Cal pH/ATC Probe, SJ/Epoxy; BNC/8-Pin DIN
Representative image only.
Thermo Scientific Mfr # 5107BNMD – Item # WW-05719-06
Easy-to-clean and accurate
  • For use in the lab, plant, or field
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Reference JunctionCeramic
  • Min pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature Accuracy±1.0°C
  • Temperature CompensationYes
  • ATC Connector8-pin mini-DIN
  • ConnectorBNC
  • Cable Length (m)1
  • DescriptionNo-Cal pH/ATC Probe, SJ/Epoxy; BNC/8-Pin DIN
  • Warranty1 year
1 Year Warranty
Key Features
  • For use in the lab, plant, or field
  • 1 year warranty
More About this Item
Make quick, accurate measurements with no calibration required using Thermo Scientific™ Orion™ No-Cal™ pH/ATC Combination Electrodes.