Texas Electronics CWS-002 Weather Station Mast Cross Arm
Representative image only.
Texas Electronics Mfr # CWS-002 – Item # WW-99535-74
Securely mount your Independent Weather Sensors
  • Finished with backed enamel coating for enhanced durability
  • Quick-release wiring harness for easy installation and maintenance
  • Constructed of aluminum to secure weather sensors
  • 1" H by 3" W crossarm with 4' length
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionWeather Station Mast Cross Arm
  • Warranty3 year
3 Year Warranty
Key Features
  • Finished with backed enamel coating for enhanced durability
  • Quick-release wiring harness for easy installation and maintenance
  • Constructed of aluminum to secure weather sensors
  • 1" H by 3" W crossarm with 4' length
  • 3 year warranty
More About this Item
This weather station is designed to fit applications where precise weather data is required. The system utilizes the highest quality sensors and rugged mounting hardware to ensure reliable data transfer. Sensors and datalogging systems sold separately.