Testo 0628 7533 Temperature probe, immersion, for 92600 series of dataloggers
Representative image only.
Testo Mfr # 0628 7533 – Item # WW-92600-42
Durable temperature probe for Testo dataloggers
  • TC Type K temperature probe with stainless steel sleeve and 1.9 m cable
  • Temperature measurement in liquids, pastes and also in air
  • Measuring range: -58 to 401 °F (-50 to +205 °C)
  • Reaction time: 20 seconds
  • Probe dimensions are 6 mm diameter by 40 mm length
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTemperature probe, immersion, for 92600 series of dataloggers
  • Warranty2 year
Two year warranty
Key Features
  • TC Type K temperature probe with stainless steel sleeve and 1.9 m cable
  • Temperature measurement in liquids, pastes and also in air
  • Measuring range: -58 to 401 °F (-50 to +205 °C)
  • Reaction time: 20 seconds
  • Probe dimensions are 6 mm diameter by 40 mm length
  • 2 year warranty
More About this Item
This thermocouple (TC) temperature probe with flattened tip can be used for measurements in liquids, pastes and also for measurements in air.
The temperature probe features an acid-resistant, stainless steel sleeve and is suitable for measurements in the food industry.
With its 1.9 m cable, the temperature probe with stainless steel sleeve can also be used at measuring locations which are not in close proximity to the Wi-Fi data logger.
This Type K, Class 2 thermocouple probe has a standardized accuracy of 4.5 (±2.5 °C).
This probe's response time t99 (time it takes for the probe to register 99% of the jump in temperature) of 20 seconds refers to measurements in moving water at 140 °F (+60 °C). This response time is extended when, for example, measurements are taken in still liquid, pastes or in air.
This probe can be connected to the testo Saveris 2-T3 Wi-Fi data logger, sold separately.