Testo 0554 0493 Reflective Tape for 460 Compact Photo Tachometer; 5 pieces
Representative image only.
Testo Mfr # 0554 0493 – Item # WW-54501-01
Take rpm measurements with precision accuracy
  • Perform preventive maintenance inspections from a safe distance
  • Compact design is perfect for any location
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • RPM Accuracy (rpm)±0.02% of reading + 1 digit
  • DescriptionReflective Tape for 460 Compact Photo Tachometer; 5 pieces
  • Warranty1 year
1 Year Warranty
Key Features
  • Perform preventive maintenance inspections from a safe distance
  • Compact design is perfect for any location
  • 1 year warranty
More About this Item
This noncontact tachometer provides optical rpm measurements using a built-in LED sighting design.
Unit is ideal for measuring the speed of motors, fans, rpm of rollers, monitoring turbines, gear movement and engagement, and cam and spindle motion. Tachometer provides min/max values and displays measurements on the large, backlit LCD that is easily read in dim or poorly lit environments. The ergonomic design fits comfortably in your hand making this tachometer simple to operate.