Temperature Data Logger Download and Analysis Software
Representative image only.
Item # WW-18005-04
Quality accessories for Traceable® Temperature Data Loggers
  • Download and analyze data, or program individual units
PRODUCT OPTIONS
₹29,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible With18005-00, 18005-01, 18005-03, 18005-05
  • DescriptionTemperature Data Logger Download and Analysis Software
1 Year Warranty
Key Features
  • Download and analyze data, or program individual units
More About this Item
The software is required to download and analyze data, or program individual units. One copy of the software may be used for multiple data loggers. Software allows the user to update settings including: individual unit name/description, time zone settings, daylight savings time on/off, unit of measure (°F or °C), start delay, recording duration/sampling interval, alarm setting, quarantine value and its alarm, and user stop key enabling.
USB interface and cable (required) is sold separately.