Tektronix Service Solutions DI10103 Intrinsically Safe high speed anemometer
Representative image only.
Tektronix Service Solutions Mfr # A/4-4" – Item # WW-32044-81
Accurate measurements in a rugged unit
  • Intrinsically safe 4" high speed mechanical anemometer
  • Accurately measures accumulated air movement and speed
  • 1 year warranty
PRODUCT OPTIONS
₹207,000.00INR / Each
₹33,800.00 - ₹41,900.00INR / Each
for an additional
₹33,800.00INR / Each
for an additional
₹41,900.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Max Air Velocity (ft/min)10000
  • Min Air Velocity (ft/min)200
  • DescriptionIntrinsically Safe high speed anemometer
  • Warranty1 year
1 Year Warranty
Key Features
  • Intrinsically safe 4" high speed mechanical anemometer
  • Accurately measures accumulated air movement and speed
  • 1 year warranty
More About this Item
This first-class hand anemometer has wide appeal, providing the simplest and most reliable measurement of air movement. Rugged and sensitive, yet compact and easy to operate. Used by government departments, mines, industrial plants, laboratories, colleges, contractors, and more. All Davis Instruments anemometers are completely U.S.A. manufactured, assembled and calibrated, and have long life expectancy with lasting accuracy.