Cole-Parmer Standard Basket for 5 L Industrial Fluidized Sand Bath, Steel
Representative image only.
Cole-Parmer Item # WW-01184-64
  • Durable steel mesh construction
  • Safely hold samples away from heating elements
  • Easily retrieve samples from the bath
  • 3 year warranty
  • Previous part number FA624
PRODUCT OPTIONS
₹94,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialSteel
  • Compatible WithIndustrial fluidized sand bath 01184-10
  • DescriptionStandard Basket for 5 L Industrial Fluidized Sand Bath, Steel
  • Warranty3 Years
3 Year Warranty
Key Features
  • Durable steel mesh construction
  • Safely hold samples away from heating elements
  • Easily retrieve samples from the bath
  • 3 year warranty
  • Previous part number FA624
More About this Item
Parts and accessories to meet your application needs and keep your sand bath operating dependably.