Cole-Parmer Essentials Hollow Polypropylene (PP) Ball Bath Covers, 10 mm, 5000/pk
Representative image only.
Cole-Parmer Essentials Item # WW-06821-12
Reduce vapor and heat loss by 75% with one layer of balls
PRODUCT OPTIONS
₹106,000.00INR / Pkg of 5000
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHollow Polypropylene (PP) Ball Bath Covers, 10 mm, 5000/pk
More About this Item
Polypropylene balls form a tight cover over liquids to prevent change in temperature and reduce evaporation. Use instead of a bath lid with any of the baths listed in the section.